Skip to Content
Merck
All Photos(1)

Key Documents

111139

Sigma-Aldrich

3-Methyl-2-pentanol

99%

Synonym(s):

sec-Butyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.421 (lit.)

bp

131-134 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

CCC(C)C(C)O

InChI

1S/C6H14O/c1-4-5(2)6(3)7/h5-7H,4H2,1-3H3

InChI key

ZXNBBWHRUSXUFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-2-pentanol is a metabolite found in the urine of rats exposed to controlled concentration of 3-methylpentane[1]. The metabolites can be analyzed by the gas chromatographic method[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Frontali et al.
Clinical toxicology, 18(12), 1357-1367 (1981-12-01)
Rats were intermittently exposed (9 to 10 h/d, 5 to 6 d/week) to controlled concentrations of single analytical grad solvents in ambient air. After periods ranging from 7 to 30 weeks the animals were perfused with glutaraldehyde and samples of
L Perbellini et al.
International archives of occupational and environmental health, 48(1), 99-106 (1981-01-01)
A gas chromatographic method for analyzing the urinary metabolites of n-hexane (2-hexanol, 2,5-hexanedione, 2,5-dimethylfuran and gamma-valerolactone), of 2-methylpentane (2-methyl-2-pentanol), of 3-methylpentane (3-methyl-2-pentanol), and of cyclohexane (cyclohexanol) was developed. Processing of urine and the gas chromatographic conditions are described. The recovery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service