Skip to Content
Merck
All Photos(1)

Key Documents

AC7300012911

Advanced Chemical Intermediates Ltd.

Ethyl 6-chloro-5-cyano-2-phenylnicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
286.72
MDL number:

Iupac Name

ethyl 6-chloro-5-cyano-2-phenylnicotinate

Inchi Code

1S/C15H11ClN2O2/c1-2-20-15(19)12-8-11(9-17)14(16)18-13(12)10-6-4-3-5-7-10/h3-8H,2H2,1H3

InChI key

NXOAKQSGNPAYSU-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Product Link

https://www.acints.com/products/ACI-NI-00058

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/ACI-NI-00058


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library