AABH14DD978E
AA BLOCKS, INC.
propan-2-yl 3-cyano-4-(propan-2-yloxy)benzoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
isopropyl 3-cyano-4-isopropoxybenzoate
Inchi Code
1S/C14H17NO3/c1-9(2)17-13-6-5-11(7-12(13)8-15)14(16)18-10(3)4/h5-7,9-10H,1-4H3
InChI key
PXOPMWGRVJKKHM-UHFFFAOYSA-N
storage temp.
2~8 C
Country of Origin
US
Product Link
https://www.aablocks.com/prod/1261173-10-7
Synonym
Isopropyl3-Cyano-4-isopropoxybenzoate
MSDS
https://www.aablocks.com/downloadSds?cas=1261173-10-7
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.