![(2R,3R)-(+)-2,3-Bis(diphenylphosphino)butane](/deepweb/assets/sigmaaldrich/product/structures/262/775/fcb7a934-5526-40ad-a39d-09f851f4da85/640/fcb7a934-5526-40ad-a39d-09f851f4da85.png)
(2R,3R)-(+)-2,3-Bis(diphenylphosphino)butane
![(2R,3R)-(+)-2,3-Bis(diphenylphosphino)butane](/deepweb/assets/sigmaaldrich/product/structures/262/775/fcb7a934-5526-40ad-a39d-09f851f4da85/640/fcb7a934-5526-40ad-a39d-09f851f4da85.png)
Synonym(s):
(R,R)-Chiraphos
Empirical Formula (Hill Notation):
C28H28P2
CAS Number:
Molecular Weight:
426.47
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1