Skip to Content
Merck
All Photos(2)

Key Documents

SRE0116

Sigma-Aldrich

Salt Active Nuclease (SAN)

recombinant, expressed in Pichia pastoris, Triton-free

Synonym(s):

endonuclease, recombinant nuclease, salt active endonuclease, salt tolerant nuclease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

recombinant

expressed in Pichia pastoris

Quality Level

composition

Triton-free

packaging

pkg of 25 kU

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This Salt Active Nuclease (SAN) has been developed to offer the optimal solution for removal of nucleic acids at the near-physiological conditions used in many bioprocessing and biomanufacturing workflows. This novel, nonspecific endonuclease is active over a broad pH range and displays optimum activity at salt concentrations around 175 mM. It is recombinantly expressed in Pichia pastoris.

Application

  • Can be used directly in cell medium or harvested supernatant without buffer adjustments
  • Ideal for DNA removal directly in mammalian cell media

Features and Benefits

  • Active at physiological conditions
  • Endotoxin tested Animal origin free without buffer adjustments.

Unit Definition

One unit is defined as the amount of enzyme that causes a ΔA260 = 1.0 in 30 minutes at 37 °C in 25 mM Tris-HCl pH 7.6 (@25 °C), 2.5 mM MgCl2, 150 mM NaCl, and 50 μg/ml calf thymus DNA. 25 mM Tris-HCl pH 7.6 (@25 °C), 2.5 mM MgCl2, 150 mM NaCl, and 50 μg/ml calf thymus DNA

Reconstitution

In solution

Other Notes

  • Temperature: 25 – 50 °C
  • Salt concentration (NaCl): 50 – 400 mM, optimal: 125 – 250 mM
  • Mg2+: > 0.5 mM is required for activity, optimal: 4 - 15 mM
  • pH: 6.5 – 9.5, optimal: 7.2 - 8.7
  • Not recommended to use in presence of EDTA or urea.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service