SML3747
IM1761092 hydrochloride
≥98% (HPLC)
Synonym(s):
IM 1092 hydrochloride, IM 1761092 hydrochloride, IM-1092 hydrochloride, IM-1761092 hydrochloride, IM1092 hydrochloride, N-1-(3-chloro-4-iodo)phenethylbiguanide hydrochloride
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 2 mg/mL, clear
storage temp.
room temp
SMILES string
ClC1=CC(CCNC(NC(N)=N)=N)=CC=C1I.Cl
InChI key
NSBWTYOKPDFWFE-UHFFFAOYSA-N
Biochem/physiol Actions
Caution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service