Skip to Content
Merck
All Photos(1)

Key Documents

SML3227

Sigma-Aldrich

KI-ARv-03

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N5
CAS Number:
Molecular Weight:
259.35
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

(Powder or Lyophilized powder or film or oil)

color

colorless to brown-yellow

storage temp.

2-8°C

SMILES string

CCCC1=NC2=CC=NN2C(N[C@H]3C[C@@H](CC3)N)=C1

Biochem/physiol Actions

KI-ARv-03 is a cell penetrant, potent and selective inhibitor of CDK9 that that reduces AR-driven transcription and proliferation in prostate cancer cells. KI-ARv-03 modulates AR and AR-V7-dependent transcription in 22Rv1 cells. It exhibits potent anti-tumor activity in castration-resistant prostate cancer (CRPC) models.
Orally available, potent, ultraselective CDK9 inhibitor that exhibits potent anti-tumor activity in castration-resistant prostate cancer (CRPC) models

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Richters et al.
Cell chemical biology, 28(2), 134-147 (2020-10-22)
Castration-resistant prostate cancers (CRPCs) lose sensitivity to androgen-deprivation therapies but frequently remain dependent on oncogenic transcription driven by the androgen receptor (AR) and its splice variants. To discover modulators of AR-variant activity, we used a lysate-based small-molecule microarray assay and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service