Skip to Content
Merck
All Photos(1)

Key Documents

EP0030504216

Eppendorf® Deepwell plates, Protein LoBind, 96 wells

yellow plate, conical bottom colorless wells, capacity 1000 μL, pkg of 20 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

conical bottom colorless wells
yellow plate

packaging

pkg of 20 ea

manufacturer/tradename

Eppendorf® 0030504216

technique(s)

PCR: suitable

capacity

1000 μL

size

96 wells

working volume

1 mL

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Maximize your protein sample recovery with Eppendorf Protein LoBind Plates. Eppendorf LoBind Plates are equipped with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for protein research or sensitive testing methods in proteomics, where they enable excellent results due to less sample binding to the plate surface.

Application

  • Preparation or storage of protein, peptide or antibody samples
  • Enzymatic assays – the hydrophilic surface reduces denaturation when interacting with the vessel wall
  • Avoiding sample loss when storing virus stock solutions
  • Storage of cell suspensions

Features and Benefits

  • Eppendorf LoBind material ensures outstanding sample recovery for better test results
  • Without surface coating (e.g. silicone) to minimize the risk of sample interference
  • Batch-certified purity grade PCR clean: free from human DNA, DNase, RNase and PCR inhibitors
  • Available as reaction vessel, microplate and deepwell plate for easy scaling
  • Unique high-contrast OptiTrack matrix: up to 30% faster sample detection and fewer pipetting errors
  • RecoverMax well design: Optimized well geometry for minimized residual or dead volume and excellent mixing properties
  • Raised corrugated edges and a smooth surface ensure reliable sealing of the plates

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service