Skip to Content
Merck
All Photos(1)

Key Documents

L8

95102434, rat muscle, Myoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
rat muscle
growth mode:
Adherent
karyotype:
Not specified
morphology:
Myoblast
products:
Not specified
receptors:
Not specified

Product Name

L8, 95102434

biological source

rat muscle

growth mode

Adherent

karyotype

Not specified

morphology

Myoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Skeletal muscle myoblast

Cell Line Description

The myogenic cell line L8 was isolated by D Yaffe in 1969 by selective serial passage of myoblasts from primary rat skeletal muscle cell cultures (new born, non-in-bred Wistar rats). Subsequently they were recloned by H Blau. L8 cells differentiate into cross-striated myotubes by fusion and express several muscle-specific proteins. The myoblast population will be depleted if cells are allowed to become confluent before sub-culturing. Therefore, sub-cultivation before confluency and periodical re-cloning is necessary. The preparation of a frozen stock at early passage is recommended.

Application

Study of muscle differentiation, expression and analysis of muscle-specific proteins

Culture Medium

DMEM : Medium 199 (4:1) + 2mM Glutamine + 1% CEE (Chick Embryo extract (Seralab)) + 10% Horse Serum (HS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000 cells to 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C; sub-cultivate before confluency and reclone periodically.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service