Skip to Content
Merck
All Photos(1)

Key Documents

LC540

89031604, rat testis, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
rat testis
growth mode:
Adherent
karyotype:
2n = 42
morphology:
Epithelial
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

LC540, 89031604

biological source

rat testis

growth mode

Adherent

karyotype

2n = 42

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Fischer Leydig cell testicular tumour

Cell Line Description

A cell line from a spontaneous transplantable Leydig cell tumour in a Fischer strain rat, reported to secrete steroid, however over a 6 hour period the secretion of oestradiol and testosterone by radioimmunoassay could not be confirmed.

Application

Tumourigenicity studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service