Skip to Content
Merck
All Photos(1)

Key Documents

RPMI 1788

85112106, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

RPMI 1788, 85112106

biological source

human blood

growth mode

Suspension

karyotype

2n = 46, modal no. 45

morphology

Lymphoblast

products

Immunoglobulin M (IgM) (Lambda light chain)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian peripheral blood lymphocyte

Cell Line Description

Derived from the peripheral blood of an apparently normal 33 year old Caucasian male. The cells secrete IgM (lambda light chain). HLA = A2, Aw33, B7, B14.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 10
D13S317: 11,13
D16S539: 10,13
D5S818: 12,13
D7S820: 10,12
THO1: 6,9.3
TPOX: 8,9
vWA: 18,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-4x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 1-2x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service