Skip to Content
Merck
All Photos(1)

Key Documents

08776

Supelco

Ytterbium(III) Ionophore II

Selectophore, function tested

Synonym(s):

3-Hydroxy-2-naphthoic(2-hydroxybenzylidene)hydrazide, 3-Hydroxy-N′-[(2-hydroxyphenyl)methylene]-2-naphthydrazide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

quality

function tested

SMILES string

Oc1ccccc1\C=N/NC(=O)c2cc3ccccc3cc2O

InChI

1S/C18H14N2O3/c21-16-8-4-3-7-14(16)11-19-20-18(23)15-9-12-5-1-2-6-13(12)10-17(15)22/h1-11,21-22H,(H,20,23)/b19-11-

InChI key

XTQAGBABCDSRSN-ODLFYWEKSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Nano-level monitoring of ytterbium(III)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganjali, M.R., et al.
Sensors and Actuators B, Chemical, 114, 855-860 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service