Skip to Content
Merck
All Photos(2)

Key Documents

8.14680

Sigma-Aldrich

3,3′-Diaminodiphenyl sulfone

for synthesis

Synonym(s):

3,3′-Diaminodiphenyl sulfone, 3,3′-Sulfonyldianiline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O2S
CAS Number:
Molecular Weight:
248.30
MDL number:
UNSPSC Code:
12352116
EC Index Number:
209-967-5
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

form

solid

potency

4920 mg/kg LD50, oral (Rat)

mp

167-170 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(c2cc(ccc2)N)c1cc(ccc1)N

InChI

1S/C12H12N2O2S/c13-9-3-1-5-11(7-9)17(15,16)12-6-2-4-10(14)8-12/h1-8H,13-14H2

InChI key

LJGHYPLBDBRCRZ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 166 °C
Melting range (upper value): ≤ 170 °C
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service