Skip to Content
Merck
All Photos(1)

Key Documents

524734

Sigma-Aldrich

Phosphodiesterase 3B, His•Tag®, Human, Recombinant, S. frugiperda

Synonym(s):

PDE3B, cGMP-inhibited phosphodiesterase 3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

form

liquid

Quality Level

specific activity

≥0.5 U/μL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−70°C

General description

Full-length, recombinant, human phosphodiesterase 3B (accession no. AY459346) fused at the N-terminus to a His•Tag sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. An important component of insulin and cAMP-dependent signaling. Supplied as a partially purified preparation.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole cAMP to AMP per min at pH 7.5, 30°C.

Physical form

In 150 mM NaCl, 10 mM Tris, 0.1% NP-40, pH 8.3.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Gibson, L.C.D., et al. 2006. Eur. J. Pharmacol.538, 39.
Kitamura, T., et al. 1999. Mol. Cell. Biol.19, 6286.
Degerman, E., et al. 1997. J. Biol. Chem.272, 6823.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service