218767
Caspase-6 Inhibitor II, Cell-Permeable
The Caspase-6 Inhibitor II, Cell-Permeable controls the biological activity of Caspase-6. This small molecule/inhibitor is primarily used for Cancer applications.
Synonym(s):
Caspase-6 Inhibitor II, Cell-Permeable, VEID-CHO, Cell-permeable; sequence
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
Assay
≥97% (HPLC)
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
solubility
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
A potent, cell-permeable, and reversible inhibitor of caspase-6.The N-terminal sequence (amino acids 1-16) corresponds to the hydrophobic region of the signal peptide of Kaposi fibroblast growth factor and confers cell permeability to the peptide.
Potent, cell-permeable and reversible inhibitor of caspase-6.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
caspase-6
caspase-6
Product does not compete with ATP.
Reversible: yes
Warning
Toxicity: Standard Handling (A)
Sequence
Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Glu-Ile-Asp-CHO
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Hirata, H., et al. 1998. J. Exp. Med. 187, 587.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service