Skip to Content
Merck
All Photos(1)

Documents

778176

Sigma-Aldrich

N-Acetyl-Asp-Glu-OH-15N2

98 atom % 15N, 95% (CP)

Synonym(s):

Spaglumic acid-15N2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H1615N2O8
Molecular Weight:
306.24
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

98 atom % 15N

Assay

95% (CP)

form

solid

technique(s)

bio NMR: suitable
mass spectrometry (MS): suitable
protein expression: suitable

mass shift

M+2

storage temp.

−20°C

SMILES string

CC([15NH][C@@H](CC(O)=O)C([15NH][C@@H](CCC(O)=O)C(O)=O)=O)=O

InChI

1S/C11H16N2O8/c1-5(14)12-7(4-9(17)18)10(19)13-6(11(20)21)2-3-8(15)16/h6-7H,2-4H2,1H3,(H,12,14)(H,13,19)(H,15,16)(H,17,18)(H,20,21)/t6-,7-/m0/s1/i12+1,13+1

InChI key

OPVPGKGADVGKTG-VRXKZSBSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service