Merck
  • Endoplasmic reticulum stress mediates radiation-induced autophagy by perk-eIF2alpha in caspase-3/7-deficient cells.
Endoplasmic reticulum stress mediates radiation-induced autophagy by perk-eIF2alpha in caspase-3/7-deficient cells.
Oncogene (2010-03-30)
K W Kim, L Moretti, L R Mitchell, D K Jung, B Lu
ABSTRACT
As apoptosis defects limit efficacy of anticancer agents, autophagy has been proposed as a novel strategy for radiotherapy enhancement. We previously showed that caspase-3/7 inhibition induces autophagy and promotes radiosensitivity in vitro and in vivo. Therefore, we further investigated the mechanism by which radiation triggers autophagy in caspase-3/7-deficient cells, and found the involvement of endoplasmic reticulum (ER) stress. The ER activates a survival pathway, the unfolded protein response, which involves ER-localized transmembrane proteins such as protein kinase-like ER kinase (PERK), inositol-requiring enzyme-1 and activating transcription factor-6. In this study, we found that PERK is essential for radiation-induced autophagy and radiosensitivity in caspase-3/7 double-knockout cells. Irradiation of these cells increased expression of phosphorylated-eIF2alpha. Similar results were seen after administration of tunicamycin (TM), a well-known ER stressor. Importantly, we found that the administration of TM with radiation in MCF-7 breast cancer cells, which are lacking functional caspase-3 and relatively resistant to many anticancer agents, enhances radiation sensitivity. Our findings reveal ER stress as a novel potential mechanism of radiation-induced autophagy in caspase-3/7-deficient cells and as a potential strategy to maximize efficiency of radiation therapy in breast cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human ERN1