Merck
  • Synthesis of multisubstituted quinolines from Baylis-Hillman adducts obtained from substituted 2-chloronicotinaldehydes and their antimicrobial activity.
Synthesis of multisubstituted quinolines from Baylis-Hillman adducts obtained from substituted 2-chloronicotinaldehydes and their antimicrobial activity.
Bioorganic & medicinal chemistry (2006-03-03)
P Narender, U Srinivas, M Ravinder, B Ananda Rao, Ch Ramesh, K Harakishore, B Gangadasu, U S N Murthy, V Jayathirtha Rao
ABSTRACT
Baylis-Hillman acetates were synthesized from substituted 2-chloronicotinaldehydes and were conveniently transformed into multisubstituted quinolines and cyclopenta[g]quinolines on reaction with nitroethane or ethyl cyanoacetate via a successive S(N)2'-S(N)Ar elimination strategy. Thus, synthesized quinolines were evaluated for antimicrobial activity and found having substantial antibacterial and antifungal activity.
Sigma-Aldrich
3-Pyridinecarboxaldehyde, 98%