Merck
  • Influence of CYP2C9 Genetic Polymorphisms on the Pharmacokinetics of Losartan and Its Active Metabolite E-3174: A Systematic Review and Meta-Analysis.
Influence of CYP2C9 Genetic Polymorphisms on the Pharmacokinetics of Losartan and Its Active Metabolite E-3174: A Systematic Review and Meta-Analysis.
Journal of personalized medicine (2021-07-03)
Yoon-A Park, Yu-Bin Song, Jeong Yee, Ha-Young Yoon, Hye-Sun Gwak
ABSTRACT
This study aimed to investigate the influence of CYP2C9 genetic polymorphisms on the pharmacokinetics of losartan and its active metabolite, E-3174, through a systematic review and meta-analysis. Eight studies published before March 2021 were included in this study. We used PubMed, the Cochrane Library, EMBASE, and Web of Science, based on the Preferred Reporting Items for Systematic Reviews and Meta-Analyses (PRISMA) guidelines. The data analysis was conducted through Review Manager (RevMan), version 5.3, and R software. We found that healthy volunteers with CYP2C9*2 or *3 carriers had higher area under the curve (AUC0-∞) of losartan (mean difference (MD) 0.17 μg·h/mL; 95% confidence intervals (CI): 0.04, 0.29) and lower AUC0-∞ of E-3174 (MD -0.35 μg·h/mL; 95% CI: -0.62, -0.08) than those with CYP2C9*1/*1. Subjects with CYP2C9*2 or *3 carriers showed lower maximum concentration (Cmax) of E-3174 than those with CYP2C9*1/*1 (MD -0.13 μg/mL; 95% CI: -0.17, -0.09). For half-life, subjects with CYP2C9*2 or *3 carriers had longer half-lives of losartan and E-3174 than those with CYP2C9*1/*1 (MD 0.47 h; 95% CI: 0.32, 0.61 and MD 0.68 h; 95% CI: 0.44, 0.92, respectively). This meta-analysis suggests that the pharmacokinetics of losartan and E-3174 are associated with the CYP2C9 polymorphisms.
Sigma-Aldrich
Losartan, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Losartan potassium, ≥98% (HPLC)