
TWEEN® 80 (Polysorbate)

Synonym(s):
TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80
CAS Number:
Products
Product Category
Brand
Physical Form
Product Line
Agency
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing