G005151
GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DP
suitable for GC
About This Item
technique(s)
gas chromatography (GC): suitable
test parameters
column: Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven: 140 °C
detector: FID, 250 °C
inj. temp.: 250 °C
carrier gas: helium, 2.7 mL/min., constant flow
injection: 1 μL, 80:1 split
liner: 4 mm I.D., FocusLiner
sample: 4 mg/mL each compound in methylene chloride
peak 1: d-amphetamine (N-TFA derivative)
peak 2: l-amphetamine (N-TFA derivative)
application(s)
clinical
forensics and toxicology
pharmaceutical (small molecule)
Analysis Note
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service