Skip to Content
Merck
All Photos(1)

Documents

CRM80838

Supelco

Metalaxyl (S)-enantiomer

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-(2,6-Dimethylphenyl)-N-(2-methoxyacetyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Metalaxyl (S)-enantiomer is the S-enantiomer of the widely used acetylalanine fungicide― metalaxyl. A systemic pesticide, it targets the RNA polymerase complex I and inhibits the synthesis of ribosomal RNA. It shows protective and curative abilities after its absorption through roots, stalks, and leaves.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, metalaxyl is approved for use as a plant protection agent in the European Union. The Reg. (EU) 2017/1164 sets a maximum residue limit (MRL) of 0.7 mg/kg and 0.01 mg/kg for the presence of metalaxyl in fresh/frozen fruits and tree nuts respectively, considering other mixtures of its constituent isomers including metalaxyl-M (sum of isomers). It is also included for monitoring in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA for products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service