59407
OPTI-SEAL pump seal
for use with 655, L6000, L6200, L6200A, Hitachi 655-1080
Synonym(s):
Optimize Technologies HPLC Pump Part for Hitachi®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 1
manufacturer/tradename
Hitachi 655-1080
compatibility
for use with 655, L6000, L6200, L6200A
Looking for similar products? Visit Product Comparison Guide
General description
OPTI-SEAL pump seals are made from an inert, ultra-high molecular weight polyethylene material (UHMW-PE). Relative to conventional polytetrafluoroethylene (PTFE) seals, these seals show less particle shedding. This reduces the potential for clogged frits, resulting in the prevention of unscheduled system downtime. These seals are recommended for typical reversed phase mobile phases (aqueous to moderate levels of organic). They are not recommended for long-term use with highly organic mobile phases, such as typical normal phase solvents.
Legal Information
Hitachi is a registered trademark of Hitachi Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service