Skip to Content
Merck
All Photos(1)

Key Documents

51356-U

MXP High Pressure Valves for HPLC

PEEK, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

material

PEEK

description

MXP9900-000 2-position, 6-port

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

The MXP7900-000 valve is the standard 2-position, 6-port switching valve designed for traditional HPLC and related techniques, used for traditional sample injections or to provide dual-column functionality in your system.

The MXP9900-000 valve should be used when the application requires biocompatability.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service