Key Documents
Z740422
Tamper proof specimen containers
size 1000 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.
material
polypropylene cap
polypropylene container
packaging
pack of 100 ea
manufacturer/tradename
Simport C581-1L
size
1000 mL
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
Z740420Z740421Z740423
packaging
pack of 100 ea
packaging
pack of 100 ea
packaging
pack of 100 ea
packaging
pack of 25 ea
manufacturer/tradename
Simport C581-1L
manufacturer/tradename
Simport C581-300
manufacturer/tradename
Simport C581-500
manufacturer/tradename
Simport C581-2600
size
1000 mL
size
300 mL
size
500 mL
size
2600 mL
General description
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.