Skip to Content
Merck
All Photos(1)

Key Documents

SML3090

Sigma-Aldrich

EN4-2

≥98% (HPLC)

Synonym(s):

(E)-4-(but-2-enamidomethyl)-N-(2-(4-ethoxyphenoxy)phenyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N2O4
Molecular Weight:
430.50
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

EN4-2 is a cell penetrant, potent and selective covalent c-MYC inhibitor that targets cysteine 171 located within a predicted intrinsically disordered region of MYC. EN4-2 inhibits MYC transcriptional activity, downregulates multiple MYC transcriptional targets, reduces MYC and MAX thermal stability, and impairs tumorigenesis.
cell penetrant, potent and selective covalent c-MYC inhibitor that targets cysteine 171

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lydia Boike et al.
Cell chemical biology, 28(1), 4-13 (2020-09-24)
MYC is a major oncogenic transcriptional driver of most human cancers that has remained intractable to direct targeting because much of MYC is intrinsically disordered. Here, we have performed a cysteine-reactive covalent ligand screen to identify compounds that could disrupt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service