Skip to Content
Merck
All Photos(1)

Key Documents

SML1207

Sigma-Aldrich

GPBAR-A

≥98% (HPLC)

Synonym(s):

4-[[3,5-Bis(trifluoromethyl)phenyl]methyl]-6-(2-fluorophenyl)-4,5-dihydro-pyrido[3,2-f]-1,4-oxazepin-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15F7N2O2
CAS Number:
Molecular Weight:
484.37
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(CO1)N(CC2=CC(C(F)(F)F)=CC(C(F)(F)F)=C2)CC3=C1N=CC=C3C4=C(F)C=CC=C4

InChI

1S/C23H15F7N2O2/c24-19-4-2-1-3-17(19)16-5-6-31-21-18(16)11-32(20(33)12-34-21)10-13-7-14(22(25,26)27)9-15(8-13)23(28,29)30/h1-9H,10-12H2

InChI key

ZIXNJVGTAXRKAP-UHFFFAOYSA-N

Biochem/physiol Actions

GPBAR-A is a very potent agonist of the bile acid receptor GPBA (also known as TGR5).
GPBAR-A is a very potent agonist of the bile acid receptor GPBA (also known as TGR5). In murine primary intestinal cultures, GPBAR-A induces an elevation in intracellular cAMP and calcium levels, and stimulates secretion of GLP-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah A Goldspink et al.
Cell reports, 31(13), 107833-107833 (2020-07-02)
Glucagon-like peptide-1 (GLP-1) from intestinal L-cells stimulates insulin secretion and reduces appetite after food ingestion, and it is the basis for drugs against type-2 diabetes and obesity. Drugs targeting L- and other enteroendocrine cells are under development, with the aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service