All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C79H124N24O24
Molecular Weight:
1793.98
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
Product Name
SNP-1,
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥72%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
His-Ser-Gln-Ile-Ser-Gly-Lys-Tyr-Gln-Arg-Tyr-Leu-Lys-Asp-Ala
Application
The peptide SNP-1 (HSQISGKYQRYLKDA) is an inhibitor of ecto-ADP-ribosyl-cyclase activity of bone marrow stromal cell antigen-1(BST-1).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service