RAB0339
Human LIF ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type serum
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 8.23-6000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... LIF(3976)
Related Categories
General description
Human leukemia inhibitory factor (LIF) Enzyme-Linked Immunosorbent Assay (ELISA) kit is an in vitro assay for the quantitative measurement of human LIF in biological samples, such as serum, plasma, cell culture supernatants, urine, and/or cell and tissue lysates. LIF protein is a monomeric glycoprotein and a member of the interleukin-6 (IL-6) family of cytokines. This protein acts in an autocrine and paracrine manner. LIF participates in several biological processes like an inflammatory response, embryonic implantation, cancer progression, differentiation of leukemia cell, neuronal development, and stem cell self-renewal.
Immunogen
Recombinant Human LIF
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xuetian Yue et al.
Cancer cell & microenvironment, 2(3) (2016-01-26)
Leukemia inhibitory factor (LIF), a secreted cytokine, plays an important role in a wide array of biological processes including inducing differentiation of leukemia cell, inflammatory response, neuronal development, embryonic implantation, stem cell self-renewal and cancer progression, etc. LIF exerts its
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service