CLS356400
Corning® BioCoat™ Collagen I Multiwell Plate
well size 6 wells, flat bottom clear wells, lid
Synonym(s):
Cell Culture Plate, plastic coated cell culture plate
About This Item
Recommended Products
material
flat bottom clear wells
sterility
sterile
feature
lid
packaging
pkg of 50 ea (10 packs of 50)
manufacturer/tradename
Corning 356400
size
6 well
well size
6 wells
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service