Skip to Content
Merck
All Photos(1)

Key Documents

CLS3367

Corning® 96 Well Clear Polystyrene Microplate

round bottom clear, polystyrene, pkg of Individually wrapped, sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
polystyrene
round bottom clear
round wells
size:
96 wells
Pricing and availability is not currently available.

material

polystyrene
round bottom clear
round wells

sterility

sterile

feature

lid: no
skirt

manufacturer/tradename

Corning 3367

packaging

pkg of Individually wrapped

size

96 wells

well volume

330 μL

well working volume

75-200 μL

color

clear

suitability

suitable for (EIA/RIA; immunoassays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service