Skip to Content
Merck
All Photos(1)

Key Documents

92936

Supelco

Color Reference Solutions

related to Ph.Eur Color intensity testing, analytical standard, Set of colors B

Synonym(s):

Color calibration standard brown (B1 to B9)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

according to Ph. Eur.

shelf life

limited shelf life, expiry date on the label

packaging

ampule of 10 mL

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

The kit contains a set of nine brown color reference standards (identified sequentially from B1 to B9) and a 1% hydrogen chloride solution in water. The solutions are produced from three primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate pentahydrate, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

Application

They are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red color range.

Features and Benefits

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 10 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Components

This is a set of the following color reference solutions according to PhEur:

  • Colour Reference Solution B1 - 10mL ampule
  • Colour Reference Solution B2 - 10mL ampule
  • Colour Reference Solution B3 - 10mL ampule
  • Colour Reference Solution B4 - 10mL ampule
  • Colour Reference Solution B5 - 10mL ampule
  • Colour Reference Solution B6 - 10mL ampule
  • Colour Reference Solution B7 - 10mL ampule
  • Colour Reference Solution B8 - 10mL ampule
  • Colour Reference Solution B9 - 10mL ampule
  • Blind sample : Hydrochloric acid solution, 1% in water - 10mL ampule

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. カタログ番号92936-1SET-F 色度参照用液 欧州薬局方準拠は、EP2.2.2のreference solutions Bに相当すると認識しています。EP2.2.2 MethodⅡの場合、reference solutionsは使用時に調製することになっていますが、当製品はアンプルで密閉されているため、MethodⅡで使用することができるでしょうか?

    1 answer
    1. Yes, this product can be used in Method II.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service