Skip to Content
Merck
All Photos(1)

Key Documents

05851

Supelco

4-Aminoquinaldine

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym(s):

2-Methyl-4-quinolinamine, 4-Amino-2-methylquinoline, 4-Quinaldinamine, NSC 60281

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
Beilstein:
118323
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

analyte chemical class(es)

amino acids, nucleotide phosphates, organic acids

technique(s)

MALDI-MS: suitable

bp

333 °C (lit.)

mp

162-166 °C (lit.)

cation traces

Ba: ≤5.0 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5.0 mg/kg
Co: ≤5.0 mg/kg
Cr: ≤5.0 mg/kg
Cu: ≤5.0 mg/kg
Fe: ≤100 mg/kg
K: ≤20 mg/kg
Mg: ≤5.0 mg/kg
Mn: ≤5.0 mg/kg
Na: ≤50 mg/kg
Ni: ≤5.0 mg/kg
Pb: ≤5.0 mg/kg
Zn: ≤5.0 mg/kg

SMILES string

Cc1cc(N)c2ccccc2n1

InChI

1S/C10H10N2/c1-7-6-9(11)8-4-2-3-5-10(8)12-7/h2-6H,1H3,(H2,11,12)

InChI key

COCFIBRMFPWUDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as Maldi-Matrix material

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan R Fagerer et al.
European journal of mass spectrometry (Chichester, England), 19(1), 39-47 (2013-07-12)
Matrix-assisted laser desorption/ionization (MALDI) has been shown to be highly sensitive for analyzing low-mass compounds such as metabolites if the right matrix is used. 9-aminoacridine (9AA) is the most commonly employed matrix for negative mode MALDI-MS in metabolomics. However, matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service