Skip to Content
Merck
All Photos(2)

Key Documents

HPWP14250

Millipore

PES Membrane Filter, 0.45 μm Pore Size

Millipore Express®, filter diam. 142 mm, hydrophilic

Synonym(s):

Millipore Express PLUS Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

plain filter
polyethersulfone membrane
white filter

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore Express®
Millipore

parameter

23 mL/min water flow rate at 690 mbar (10 psi)

diam.

142 mm

filter diam.

142 mm

thickness

130-155 μm

impurities

≤0.5 EU/mL (LAL test, Aqueous extraction)

pore size

0.45 μm pore size

bubble point

≥690 mbar, air with water at 23 °C

shipped in

ambient

General description

Filter Code: HPWP
Filter Type: Screen filter
The Millipore Express PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features & Benefit:
•Ultra fast filtration and high throughput shorten process time
•Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications

Applications:
Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration

Application

  • Buffer filtration
  • Tissue culture media filtration
  • Sample filtration for PFAS analytical testing (LC-MS)

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MILLIPORE EXPRESS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service