Skip to Content
Merck
All Photos(1)

Key Documents

8.51066

Sigma-Aldrich

mono-Fmoc 1,6-diaminohexane hydrochloride

Novabiochem®

Synonym(s):

mono-Fmoc 1,6-diaminohexane hydrochloride, Fmoc-NH(CH 2) 6NH 2 · HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O2 · xHCl
CAS Number:
Molecular Weight:
338.44 (free base basis)
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reagent type: spacer

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

[Cl-].[N+H3]CCCCCCNC(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C21H26N2O2.ClH/c22-13-7-1-2-8-14-23-21(24)25-15-20-18-11-5-3-9-16(18)17-10-4-6-12-19(17)20;/h3-6,9-12,20H,1-2,7-8,13-15,22H2,(H,23,24);1H

InChI key

SKDQIWLYPJOUMY-UHFFFAOYSA-N

General description

Diaminohexane monoprotected with base-labile Fmoc protecting group.

Linkage

Replaces: 01-63-0070

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA2)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service