Skip to Content
Merck
All Photos(2)

Key Documents

8.14113

Sigma-Aldrich

(-)-Menthol

for synthesis

Synonym(s):

(-)-Menthol, 2-Isopropyl-5-methylcyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
MDL number:
UNSPSC Code:
12352104
EC Index Number:
218-690-9
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (GC)

potency

3300 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

216 °C/1013 hPa

mp

41-44 °C

transition temp

flash point 101 °C

solubility

490 mg/L

density

0.890 g/cm3 at 20 °C

bulk density

450 kg/m3

storage temp.

2-30°C

SMILES string

O[C@H]1[C@@H](CC[C@H](C1)C)C(C)C

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 41 °C
Melting range (upper value): ≤ 44 °C
Spec. rotation [α²0/D (c=10 in ethanol): -50.0 - -48.0 °
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service