Skip to Content
Merck
All Photos(1)

Key Documents

8.05901

Sigma-Aldrich

Methyl vinyl ketone

(stabilised) for synthesis

Synonym(s):

Methyl vinyl ketone, 1-Buten-3-one, MVK, Vinyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
70.09
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-160-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

100 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

491 °C

potency

23.1 mg/kg LD50, oral (Rat)
35 mg/kg LD50, skin (Rat)

expl. lim.

2.1-15.6 % (v/v)

bp

80-81 °C/1013 hPa

transition temp

flash point -7 °C

density

0.85 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

O=C(C)C=C

InChI

1S/C4H6O/c1-3-4(2)5/h3H,1H2,2H3

InChI key

FUSUHKVFWTUUBE-UHFFFAOYSA-N

General description

Methyl vinyl ketone (MVK) is an organic compound that belongs to the class of enones. It is a useful synthetic reagent in organic synthesis for the preparation of various organic compounds. MVK is a 3-oxobutyl synthon, which undergoes a 1,4 conjugate addition (Michael addition) reaction. MVK is susceptible to anionic polymerization, therefore it is typically stored with hydroquinone, which inhibits polymerization.

Application

Methyl vinyl ketone can be used as a reagent to synthesize:
  • Isocoumarins by a Pd-catalyzed regioselective cyclization reaction with o-alkynylbenzoates in the presence of LiCl.      
  • Furan and pyrrole derivatives by Rh-catalyzed 1,4-carbonylative addition reaction with arylboronic acids.      
  • Poly(p-hydroxystyrene) compounds via radical copolymerization with p-hydroxystyrene derivatives.     
  • Aryl-thiocyanatobutan-2-one derivatives by reacting with aryldiazonium salts in the presence of copper(II) catalysts.

Analysis Note

Assay (GC, area%): ≥ 93.5 % (a/a)
Density (20°C/ 4°C): 0.850 - 0.860
Water (GC, area%): ≤ 5.0 % (a/a)
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F

Flash Point(C)

-7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service