Skip to Content
Merck
All Photos(1)

Key Documents

5.05875

Sigma-Aldrich

(+)-Bicuculline

Synonym(s):

(+)-Bicuculline, GABA A Receptor Antagonist, Bicuculline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17NO6
CAS Number:
Molecular Weight:
367.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

740 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

N1([C@@H](c5c(cc6c(c5)OCO6)CC1)[C@@H]2OC(=O)c3c4c(ccc32)OCO4)C

InChI

1S/C20H17NO6/c1-21-5-4-10-6-14-15(25-8-24-14)7-12(10)17(21)18-11-2-3-13-19(26-9-23-13)16(11)20(22)27-18/h2-3,6-7,17-18H,4-5,8-9H2,1H3/t17-,18+/m0/s1

InChI key

IYGYMKDQCDOMRE-ZWKOTPCHSA-N

General description

A potent allosteric antagonist of vertebrate GABAA receptors. Blocks GABAA-induced chloride currents in cultured neurons (IC50 = 740 nM). Shown to increase blood pressure in spontaneously hypertensive rats and cause convulsion in experimental animals. Also, increases the amplitude of spontaneous colonic contractions.

Warning

Toxicity: Toxic (F)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Ueno, S., et al. 1997. J. Neurosci.17, 625.
Uchida, I., et al. 1996. Eur. J. Pharmacol.307, 89.
Minocha, A., and Galligan, J.J. 1993. Eur. J. Pharmacol.230, 187.
Robert K.A., et al. 1993. Cardiovasc. Pharmacol.21, 156.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service