Skip to Content
Merck
All Photos(1)

Key Documents

208922-M

Millipore

CaMKII Inhibitor, CK59

The CaMKII Inhibitor, CK59, also referenced under CAS 140651-18-9, controls the biological activity of CaMKII. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

CaMKII Inhibitor, CK59, 2-(2-Hydroxyethylamino)-6-aminohexylcarbamic acid tert-butyl ester-9-isopropylpurine, CK59

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H37N7O3
CAS Number:
Molecular Weight:
435.56
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1nc(c(c1N)N=Nc2ccc(cc2)O)N

InChI

1S/C9H10N6O/c10-8-7(9(11)15-14-8)13-12-5-1-3-6(16)4-2-5/h1-4,16H,(H5,10,11,14,15)

InChI key

AYZRKFOEZQBUEA-UHFFFAOYSA-N

General description

A cell-permeable Olomoucine (Cat. Nos. 495620 & 495624) analog that acts as a CaMKII inhibitor (IC50<10 µM) and inhibits insulin-stimulated glucose uptake (IC50 ≤5 µM) and GLUT4 membrane translocation (IC50 = 100 µM) in 3T3-L1 adipocytes.
A cell-permeable Olomoucine analog that acts as a CaMKII inhibitor (IC50<10 M) and inhibits insulin-stimulated glucose uptake (IC50 ≤5 M) and GLUT4 membrane translocation (IC50 = 100 M) in 3T3-L1 adipocytes.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot, and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Konstantopoulos, N., et al. 2007. Endocrinology, 148, 374.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service