Skip to Content
Merck
All Photos(2)

Key Documents

1.10007

Supelco

Nitrite Test Strips, colorimetric

greener alternative

2-80 mg/L (NO₂⁻), MQuant®

Synonym(s):

Nitrite chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Nitrite Test, colorimetric, 2-80 mg/L (NO2-), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

2-80 mg/L (NO2-)

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 2 - 5 - 10 - 20 - 40 - 80 mg/L (NO2-), MQuant®

Nitrite is a pollution indicator. Therefore there are interesting application possibilities in the analysis of drinking water, waste water, industrial water and aquarium water. In food analysis, the determination of nitrite in meat products, pickling solutions, brine and deep frozen spinach is particularly important. On the MQuant® Nitrite test strips, nitrite ions react with an aromatic amine to form a diazonium salt, which in turn reacts with N-(1-naphthyl)ethylenediamine to form a red-violet azo dye. The nitrite concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the nitrite concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Nitrite test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Clinical and laboratory features of urinary tract infections in young infants: Discusses diagnostic challenges and characteristics of urinary tract infections in infants, stressing the importance of accurate detection to guide treatment (Lo et al., 2018).

  • Differentiating Kawasaki disease from urinary tract infection in febrile children with pyuria and C-reactive protein elevation: Focuses on differential diagnosis in pediatric care, which can influence the choice of nitrite tests for accurate and timely diagnosis (Han and Lee, 2018).

  • Early features of Kawasaki disease with pyuria in febrile infants younger than 6 months: Addresses the overlap of urinary symptoms in Kawasaki disease, underscoring the utility of nitrite tests in complex cases (Yoon et al., 2018).

  • Current practice on the management of pre-operative urine dipstick results in women undergoing gynaecological surgery in Wales: Reviews procedures related to pre-surgical care in gynaecology, indicating broader applications of nitrite testing in hospital protocols (Walker et al., 2019).

Linkage

Replaces: 10007-1; 10007

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service