Skip to Content
Merck
All Photos(1)

Key Documents

M-103

Supelco

Modafinil-d10 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H5NO2SD10
CAS Number:
Molecular Weight:
283.41
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(CS(C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H])=O)=O

InChI

1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

YFGHCGITMMYXAQ-LHNTUAQVSA-N

General description

A new internal standard suitable for numerous GC/MS and LC/MS applications from clinical toxicology testing and forensic analysis to urine drug testing and isotope dilution methods. Modafinil is an analeptic drug sold under the brand names Provigil® or Modavigil® and approved by the US FDA for the treatment of narcolepsy, shift work sleep disorder, and excessive daytime sleepiness. A well known nootropic, the drug is often used as a performance-enhancer in academics and sports.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Modavigil is a registered trademark of Cephalon, Inc.
Provigil is a registered trademark of Cephalon, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service