Key Documents
Z758108
Wash-N-Dry Slide Rack
pack of 1 ea
Synonym(s):
slide rack trays, slide racks
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
Pricing and availability is not currently available.
material
glass filled polypropylene
feature
autoclavable
Holds 2 large coverslips and microscope slides ranging in width from 22 to 26mm
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech WSDR-2000
General description
MAXIMUM CIRCULATION - Offset design allows liquid and air to completely flow around the entire slide ensuring cleanliness and faster drying times
“HANDS OFF DESIGN” - Contacts a minimal amount of slide surface area permitting solutions and air to affect entire slide
HOLDS 12 SLIDES - Securely holds slides for convenient cleaning, treating, staining and drying. Fits standard slide sizes including 25 mm and 1 in.
LABORATORY FRIENDLY - Utilizes chemically and thermally resistant glass infused polypropylene. Autoclavable!
CREATE A SYSTEM - Use in conjunction with a 200 mL tall form beaker (No. 1060) to efficiently use solutions
Legal Information
Wash-N-Dry is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)