Z207179
Broken-glass disposal boxes
Floor
Synonym(s):
floor standing waste glass box, laboratory waste glass box
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.35
material
cardboard
feature
Holds broken glass
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art F24653-0001
availability
not available in EU
W × L × H
12 in. × 12 in. × 27 in.
Related Categories
General description
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full, safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English, French, Spanish, and German). Shipped flat.
Choose from one of the most recent versions:
Certificates of Analysis (COA)