Key Documents
Z118346
Cotton blend glove liners
size M/L, light weight
Synonym(s):
Cotton Glove Liners, resuable fabric glove
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.
material
(100%Cotton)
white gloves
size
M/L
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 12 pair
manufacturer/tradename
Sigma-Aldrich
L
9 in.
General description
Use these gloves for product protection during inspection or light manufacturing. Also ideal for use as a liner underneath outer gloves. Reversible pattern allows glove to be worn on either hand. Lightweight styles feature unhemmed cuffs.
Choose from one of the most recent versions:
Certificates of Analysis (COA)