Key Documents
SYNS560004
Synthware 180 degree high vacuum valve
size 0-4 mm
Synonym(s):
Synthware S560004,
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S560004
Related Categories
General description
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE shaft has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat . For replacement Viton® O-ring Kit, see R7010,or R108110.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)