Skip to Content
Merck
All Photos(1)

Key Documents

SYNF973014M

Synthware filter funnel with glass stopcocks

60 mL, porosity: medium

Synonym(s):

Synthware F973014M, fritted disc funnel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.51

material

borosilicate glass

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F973014M

capacity

60 mL
60 mL

pore size

10-15 μm

Looking for similar products? Visit Product Comparison Guide

General description

With two 2 mm glass stopcocks

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service