DWK73790-50
KIMBLE® centrifuge tube
with plain rim finish, conical bottom clear glass, capacity (50 mL), without cap, non-sterile
About This Item
Recommended Products
material
conical bottom clear glass
conical bottom
transparent borosilicate glass tube
sterility
non-sterile
packaging
case of 72 ea shrink wrapped in modular tray
manufacturer/tradename
Kimble® 73790-50
capacity
(50 mL)
fitting
neck (Pushfit)
Related Categories
General description
- Conical bottom
- The 5, 10, 15 sizes have a finished top for a snap-cap closure (not supplied)
- The 50mL size has a tooled top
- Shrink wrapped in modular trays to keep them clean and safe in transmit
- Manufactured from 51 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class B requirements
- These tubes are suitable for tissue culture work and general bacteriological use.
- Supplied without caps
- No marking spot
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)