Skip to Content
Merck
All Photos(1)

Key Documents

631280

Sigma-Aldrich

4-tert-Butylbenzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N
CAS Number:
Molecular Weight:
163.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

bp

235-236 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)(C)c1ccc(CN)cc1

InChI

1S/C11H17N/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7H,8,12H2,1-3H3

InChI key

MPWSRGAWRAYBJK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesc Yraola et al.
Journal of medicinal chemistry, 49(21), 6197-6208 (2006-10-13)
Structure activity relationships for semicarbazide-sensitive amine oxidase/vascular adhesion protein-1 (SSAO/VAP-1) were studied using a library of arylalkylamine substrates, with the aim of contributing to the discovery of more efficient SSAO substrates. Experimental data were contrasted with computational docking studies, thereby

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service