All Photos(1)
About This Item
Linear Formula:
124Xe
CAS Number:
Molecular Weight:
123.91
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:
Recommended Products
isotopic purity
99.9 atom %
form
gas
mass shift
M-8
SMILES string
[124Xe]
InChI
1S/Xe/i1-7
InChI key
FHNFHKCVQCLJFQ-NOHWODKXSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service