Skip to Content
Merck
All Photos(1)

Documents

17108

Sigma-Aldrich

5-Methyl-2-pyridinesulfonamide

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2S
CAS Number:
Molecular Weight:
172.20
Beilstein:
389057
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

solid

SMILES string

Cc1ccc(nc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c1-5-2-3-6(8-4-5)11(7,9)10/h2-4H,1H3,(H2,7,9,10)

InChI key

HXURYNQXMBRJLU-UHFFFAOYSA-N

Application

5-Methyl-2-pyridinesulfonamide was used to develop a copper-catalyzed aziridination protocol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoon Han et al.
Organic letters, 6(22), 4109-4112 (2004-10-22)
[reaction: see text] We have developed, on the basis of a chelation-strategy, an efficient copper-catalyzed aziridination protocol with the use of 5-methyl-2-pyridinesulfonamide and PhI(OAc)(2). The reaction proceeds smoothly under mild conditions to give aziridines in moderate to good yields in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service